3,6-Dibromo-9-(2-ethylhexyl)carbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 3,6-Dibromo-9-(2-ethylhexyl)-9H-carbazole

D4830-1G D4830-5G
TCD4830-1GEA 127 USD
TCD4830-1G TCD4830-5G
3,6-Dibromo-9-(2-ethylhexyl)carbazole ≥98.0% (by GC)
3,6-Dibromo-9-(2-ethylhexyl)carbazole
Formula: C₂₀H₂₃Br₂N
MW: 437.22 g/mol
MDL Number: MFCD18252907
CAS Number: 173063-52-0

Order Now

Specification Test Results

Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR