1,8-Dibromonaphthalene ≥98.0% (by GC)

Supplier: TCI America
Warning

D4656-1G D4656-5G
TCD4656-1GEA 55 USD
TCD4656-1G 76219-052
1,8-Dibromonaphthalene ≥98.0% (by GC)
1,8-Dibromonaphthalene
Formula: C₁₀H₆Br₂
MW: 285.97 g/mol
Boiling Pt: 140°C
MDL Number: MFCD00183574
CAS Number: 17135-74-9

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 108.0 to 112.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR