1-Ethynyl-3,5-dimethoxybenzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 3,5-Dimethoxyphenylacetylene

E1175-1G E1175-200MG
TCE1175-1GEA 200.9 USD
TCE1175-1G TCE1175-200MG
1-Ethynyl-3,5-dimethoxybenzene ≥98.0% (by GC)
1-Ethynyl-3,5-dimethoxybenzene
Formula: C₁₀H₁₀O₂
MDL Number: MFCD03839985
CAS Number: 171290-52-1

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 44.0 to 48.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR