3-Bromo-5-ethoxypyridine ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B5000-1G B5000-5G
TCB5000-1GEA 82 USD
TCB5000-1G TCB5000-5G
3-Bromo-5-ethoxypyridine ≥97.0% (by GC, titration analysis)
3-Bromo-5-ethoxypyridine
Formula: C₇H₈BrNO
MW: 202.05 g/mol
Boiling Pt: 240°C
Melting Pt: 9°C
MDL Number: MFCD00234116
CAS Number: 17117-17-8

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR