10-Bromo-7H-benzo[c]carbazole ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
B4883-1G
B4883-5G
TCB4883-1GEA
99.4
USD
TCB4883-1G
TCB4883-5G
10-Bromo-7H-benzo[c]carbazole ≥98.0% (by GC)
10-Bromo-7H-benzo[c]carbazole
Formula:
C₁₆H₁₀BrN MW: 296.17 g/mol |
MDL Number:
MFCD22054961 CAS Number: 1698-16-4 |
Specification Test Results
Appearance | White to Light gray to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 127.0 to 131.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...