2-Hydroxy-3-phenoxypropyl acrylate ≥85.0% (by GC) stabilized

Supplier: TCI America


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

H1614-25G H1614-500G
TCH1614-25GEA 39 USD
TCH1614-25G TCH1614-500G
2-Hydroxy-3-phenoxypropyl acrylate ≥85.0% (by GC) stabilized
2-Hydroxy-3-phenoxypropyl acrylate
Formula: C₁₂H₁₄O₄
Flash Pt: 89°C
MDL Number: MFCD00066300
CAS Number: 16969-10-1
EINECS: 241-045-8

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 85.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR