4-Methoxy-trans-stilbene ≥98.0% (by GC)

Supplier: TCI America

M2964-1G M2964-5G
TCM2964-1GEA 169 USD
TCM2964-1G TCM2964-5G
4-Methoxy-trans-stilbene ≥98.0% (by GC)
4-Methoxy-trans-stilbene
Formula: C₁₅H₁₄O
MW: 210.28 g/mol
Boiling Pt: 126°C
MDL Number: MFCD00017178
CAS Number: 1694-19-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 134.0 to 138.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR