2-([1,1'-Biphenyl]-3-yl)-4-chloro-6-phenyl-1,3,5-triazine ≥98.0% (by GC)

Supplier: TCI America
Warning

B5803-1G B5803-5G
TCB5803-1GEA 96 USD
TCB5803-1G TCB5803-5G
2-([1,1'-Biphenyl]-3-yl)-4-chloro-6-phenyl-1,3,5-triazine ≥98.0% (by GC)
2-([1,1'-Biphenyl]-3-yl)-4-chloro-6-phenyl-1,3,5-triazine
Formula: C₂₁H₁₄ClN₃
MW: 343.81 g/mol
MDL Number: MFCD30536776
CAS Number: 1689576-03-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 139.0 to 143.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR