3-Hydroxyisobenzofuran-1(3H)-one ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Hydroxyphthalide, 3-Hydroxy-2-benzofuran-1(3H)-one, 3-Hydroxy-1(3H)-isobenzofuranone

H1754-25G H1754-5G
TCH1754-25GEA 123 USD
TCH1754-25G TCH1754-5G
3-Hydroxyisobenzofuran-1(3H)-one ≥98.0% (by GC, titration analysis)
3-Hydroxyisobenzofuran-1(3H)-one
Formula: C₈H₆O₃
MDL Number: MFCD00956142
CAS Number: 16859-59-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 98.0 to 102.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR