Combretastatin A4 Phosphate disodium salt ≥98.0% (by HPLC)
Supplier: TCI America
C3009-100MG
C3009-20MG
TCC3009-100MGEA
634
USD
TCC3009-100MG
TCC3009-20MG
Combretastatin A4 Phosphate disodium salt ≥98.0% (by HPLC)
Combretastatin A4 Phosphate disodium salt
Formula:
C₁₈H₁₉Na₂O₈P Storage Temperature: Freezer |
MDL Number:
MFCD00943911 CAS Number: 168555-66-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...