2,6-Bis(9H-carbazol-9-yl)pyridine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 2,6-Di(9H-carbazol-9-yl)pyridine

B4961-1G B4961-200MG
76219-472EA 370 USD
76219-472 76219-474
2,6-Bis(9H-carbazol-9-yl)pyridine ≥98.0% (by HPLC, total nitrogen)
2,6-Bis(9H-carbazol-9-yl)pyridine
Formula: C₂₉H₁₉N₃
MW: 409.49 g/mol
Melting Pt: 202°C
MDL Number: MFCD28138085
CAS Number: 168127-49-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR