1-Bromocarbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1-Bromo-9H-carbazole

B4816-1G B4816-200MG
TCB4816-1GEA 93 USD
TCB4816-1G TCB4816-200MG
1-Bromocarbazole ≥98.0% (by GC)
1-Bromocarbazole
Formula: C₁₂H₈BrN
MDL Number: MFCD18450164
CAS Number: 16807-11-7

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 122.0 to 126.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR