N,N-Bis(4'-diphenylamino-4-biphenylyl)amine ≥95.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: N'-[4'-(Diphenylamino)-[1,1'-biphenyl]-4-yl]-N,N-diphenyl-[1,1'-biphenyl]-4,4'-diamine, N4-(4'-(Diphenylamino)-[1,1'-biphenyl]-4-yl)-N4',N4'-diphenyl-[1,1'-biphenyl]-4,4'-diamine

B5676-1G B5676-5G
TCB5676-1GEA 149 USD
TCB5676-1G TCB5676-5G
N,N-Bis(4'-diphenylamino-4-biphenylyl)amine ≥95.0% (by HPLC, total nitrogen)
N,N-Bis(4'-diphenylamino-4-biphenylyl)amine
Formula: C₄₈H₃₇N₃
MW: 655.85 g/mol
MDL Number: MFCD32263460
CAS Number: 167218-39-5

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 95.0 %
Melting point 209.0 to 213.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR