N-(4-Bromophenyl)-9,9-dimethyl-9H-fluoren-2-amine ≥95.0% (by HPLC, total nitrogen)

Supplier: TCI America

B6133-1G B6133-5G
TCB6133-1GEA 131 USD
TCB6133-1G TCB6133-5G
N-(4-Bromophenyl)-9,9-dimethyl-9H-fluoren-2-amine ≥95.0% (by HPLC, total nitrogen)
N-(4-Bromophenyl)-9,9-dimethyl-9H-fluoren-2-amine
Formula: C₂₁H₁₈BrN
MDL Number: MFCD32263467
CAS Number: 1644059-09-5

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 95.0 %
Melting point 122.0 to 126.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR