4,4'-Bis(decyloxy)-3-methylazobenzene ≥95.0% (by HPLC)
Supplier: TCI America
B4597-500MG
B4597-5G
TCB4597-500MGEA
173
USD
TCB4597-500MG
TCB4597-5G
4,4'-Bis(decyloxy)-3-methylazobenzene ≥95.0% (by HPLC)
4,4'-Bis(decyloxy)-3-methylazobenzene
Formula:
C₃₃H₅₂N₂O₂ MW: 508.79 g/mol |
CAS Number:
1627844-78-3 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Elemental analysis (Nitrogen) | 5.20 to 6.20 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...