Dibenzofuran-1-boronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: Dibenzo[b,d]furan-1-ylboronic acid

D5758-1G D5758-5G
TCD5758-1GEA 53 USD
TCD5758-1G TCD5758-5G
Dibenzofuran-1-boronic acid (contains varying amounts of Anhydride)
Dibenzofuran-1-boronic acid
Formula: C₁₂H₉BO₃
MDL Number: MFCD01318978
CAS Number: 162607-19-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) 95.0 to 109.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR