2',3'-Di-O-acetyl-5'-deoxy-5-fluorocytidine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2',3'-Di-O-acetyl-5'-deoxy-5-fluoro-D-cytidine, (2R,3R,4R,5R)-2-(4-Amino-5-fluoro-2-oxopyrimidin-1(2H)-yl)-5-methyltetrahydrofuran-3,4-diyl diacetate

D4969-1G D4969-25G D4969-5G
76219-084EA 63 USD
76219-084 76219-086 76219-088
2',3'-Di-O-acetyl-5'-deoxy-5-fluorocytidine ≥98.0% (by HPLC, titration analysis)
2',3'-Di-O-acetyl-5'-deoxy-5-fluorocytidine
Formula: C₁₃H₁₆FN₃O₆
MW: 329.28 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07369302
CAS Number: 161599-46-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 187.0 to 191.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR