1,3-Dibromo-5-tetradecylbenzene ≥97.0% (by GC)

Supplier: TCI America
Warning

D5258-1G D5258-5G
76219-252EA 131 USD
76219-252 76219-254
1,3-Dibromo-5-tetradecylbenzene ≥97.0% (by GC)
1,3-Dibromo-5-tetradecylbenzene
Formula: C₂₀H₃₂Br₂
CAS Number: 157761-91-6

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 35.0 to 39.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR