4,7-Dibromo-1,10-phenanthroline hydrate ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 4,7-Dibromo-1,10-phenanthroline

D4987-1G D4987-200MG
TCD4987-1GEA 462.4 USD
TCD4987-1G TCD4987-200MG
4,7-Dibromo-1,10-phenanthroline hydrate ≥98.0% (by HPLC)
4,7-Dibromo-1,10-phenanthroline hydrate
Formula: C₁₂H₆Br₂N₂·xH₂O
MW: 338.00 g/mol
Melting Pt: 236°C
MDL Number: MFCD25562940
CAS Number: 156492-30-7

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 96.0 % (calcd.on anh.substance)
Water 3.0 to 7.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR