Abiraterone Acetate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 3beta-17-(3-Pyridyl)androsta-5,16-dien-3-ol Acetate

A2891-1G A2891-200MG
TCA2891-1GEA 815 USD
TCA2891-1G TCA2891-200MG
Abiraterone Acetate ≥98.0% (by HPLC, titration analysis)
Abiraterone Acetate
Formula: C₂₆H₃₃NO₂
MW: 391.56 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00934213
CAS Number: 154229-18-2

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 145.0 to 149.0 deg-C
Specific rotation [a]20/D -40.0 to -45.0 deg (C=2,EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR