2,6-Di(2H-1,2,3-triazol-4-yl)pyridine ≥98.0% (by HPLC)
Supplier: TCI America
|
Warning
|
D4879-1G
D4879-200MG
76219-736EA
298
USD
76219-736
76219-738
2,6-Di(2H-1,2,3-triazol-4-yl)pyridine ≥98.0% (by HPLC)
2,6-Di(2H-1,2,3-triazol-4-yl)pyridine
Formula:
C₉H₇N₇ MW: 213.20 g/mol |
CAS Number:
1542234-40-1 |
Specification Test Results
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...