1,8-Bis(diphenylphosphino)naphthalene ≥97.0% (by GC)

Supplier: TCI America
Warning

B4530-1G B4530-5G
76219-402EA 185 USD
76219-402 76219-404
1,8-Bis(diphenylphosphino)naphthalene ≥97.0% (by GC)
1,8-Bis(diphenylphosphino)naphthalene
Formula: C₃₄H₂₆P₂
MDL Number: MFCD04117326
CAS Number: 153725-04-3

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR