Trimethyl(phenoxy)silane ≥97.0% (by GC)

Supplier: TCI America
Warning

T3222-25ML T3222-5ML
TCT3222-25MLEA 141 USD
TCT3222-25ML TCT3222-5ML
Trimethyl(phenoxy)silane ≥97.0% (by GC)
Trimethyl(phenoxy)silane
Formula: C₉H₁₄OSi
Boiling Pt: 211°C
Melting Pt: -55°C
Flash Pt: 41°C
MDL Number: MFCD00053666
CAS Number: 1529-17-5
EINECS: 216-211-8
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR