2-Methoxy-9,9-dimethyl-9H-fluorene ≥98.0% (by GC)
Supplier: TCI America
M3301-1G
M3301-5G
TCM3301-1GEA
55
USD
TCM3301-1G
TCM3301-5G
2-Methoxy-9,9-dimethyl-9H-fluorene ≥98.0% (by GC)
2-Methoxy-9,9-dimethyl-9H-fluorene
Formula:
C₁₆H₁₆O |
MDL Number:
MFCD32263492 CAS Number: 1514864-84-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 80.0 to 84.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...