2-Amino-4,5-dimethylbenzoic acid ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

A2850-1G A2850-200MG
TCA2850-1GEA 314.9 USD
TCA2850-1G TCA2850-200MG
2-Amino-4,5-dimethylbenzoic acid ≥97.0% (by HPLC, titration analysis)
2-Amino-4,5-dimethylbenzoic acid
Formula: C₉H₁₁NO₂
MDL Number: MFCD00156998
CAS Number: 15089-51-7
EINECS: 247-201-1

Order Now

Specification Test Results

Appearance White to Yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR