3,6-Dibromo-9-hexyl-9H-carbazole ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
D5410-1G
D5410-5G
TCD5410-1GEA
121
USD
TCD5410-1G
TCD5410-5G
3,6-Dibromo-9-hexyl-9H-carbazole ≥98.0% (by GC)
3,6-Dibromo-9-hexyl-9H-carbazole
Formula:
C₁₈H₁₉Br₂N |
MDL Number:
MFCD23099108 CAS Number: 150623-72-6 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 98.0 to 102.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...