2-([1,1'-Biphenyl]-4-yl)-4-chloro-6-phenyl-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
|
Warning
|
B5792-1G
B5792-5G
TCB5792-1GEA
62
USD
TCB5792-1G
TCB5792-5G
2-([1,1'-Biphenyl]-4-yl)-4-chloro-6-phenyl-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
2-([1,1'-Biphenyl]-4-yl)-4-chloro-6-phenyl-1,3,5-triazine
Formula:
C₂₁H₁₄ClN₃ MW: 343.81 g/mol |
MDL Number:
MFCD27979856 CAS Number: 1472062-94-4 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 163.0 to 167.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...