1,3-Dibromoazulene ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
D4662-1G
D4662-200MG
TCD4662-1GEA
536
USD
TCD4662-1G
TCD4662-200MG
1,3-Dibromoazulene ≥98.0% (by GC)
1,3-Dibromoazulene
Formula:
C₁₀H₆Br₂ |
CAS Number:
14658-95-8 |
Specification Test Results
Appearance | Dark green to Dark purple to Black powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 88.0 to 92.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...