5-Bromo-2-chloro-4-fluoroanisole ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
B5354-1G
B5354-200MG
TCB5354-1GEA
204
USD
TCB5354-1G
TCB5354-200MG
5-Bromo-2-chloro-4-fluoroanisole ≥98.0% (by GC)
5-Bromo-2-chloro-4-fluoroanisole
Formula:
C₇H₅BrClFO |
MDL Number:
MFCD07784265 CAS Number: 146447-18-9 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 72.0 to 76.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...