1,4-Di[[2,2':6',2''-terpyridin]-4'-yl]benzene ≥97.0% (by total nitrogen basis)

Supplier: TCI America
Warning

Synonyms: 1,4-Di([2,2':6',2''-terpyridin]-4'-yl)benzene

B4194-1G B4194-200MG
76219-378EA 188 USD
76219-378 76219-380
1,4-Di[[2,2':6',2''-terpyridin]-4'-yl]benzene ≥97.0% (by total nitrogen basis)
1,4-Di[[2,2':6',2''-terpyridin]-4'-yl]benzene
Formula: C₃₆H₂₄N₆
MW: 540.63 g/mol
Melting Pt: 250°C
MDL Number: MFCD02093699
CAS Number: 146406-75-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR