2,7-Diiodo-9,9-dimethylfluorene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2,7-Diiodo-9,9-dimethyl-9H-fluorene

D5032-1G D5032-5G
76219-798EA 67 USD
76219-798 76219-800
2,7-Diiodo-9,9-dimethylfluorene ≥98.0% (by GC)
2,7-Diiodo-9,9-dimethylfluorene
Formula: C₁₅H₁₂I₂
MDL Number: MFCD12827985
CAS Number: 144981-86-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 195.0 to 199.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR