2-(3'-Chlorobiphenyl-4-yl)-4,6-diphenyl-1,3,5-triazine ≥97.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

Synonyms: 2-(3'-Chloro-[1,1'-biphenyl]-4-yl)-4,6-diphenyl-1,3,5-triazine

C3163-1G C3163-200MG
TCC3163-1GEA 172.6 USD
TCC3163-1G TCC3163-200MG
2-(3'-Chlorobiphenyl-4-yl)-4,6-diphenyl-1,3,5-triazine ≥97.0% (by HPLC, total nitrogen)
2-(3'-Chlorobiphenyl-4-yl)-4,6-diphenyl-1,3,5-triazine
Formula: C₂₇H₁₈ClN₃
MW: 419.91 g/mol
MDL Number: MFCD28975107
CAS Number: 1443049-84-0

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 204.0 to 208.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR