Eprosartan Mesylate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

E1161-1G E1161-200MG
TCE1161-1GEA 303.4 USD
TCE1161-1G TCE1161-200MG
Eprosartan Mesylate ≥98.0% (by HPLC, titration analysis)
Eprosartan Mesylate
Formula: C₂₃H₂₄N₂O₄S·CH₄O₃S
MDL Number: MFCD08141807
CAS Number: 144143-96-4
Merck Index: 14,03635

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 246.0 to 250.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR