4,4'-Bis(dodecyloxy)-3-methylazobenzene ≥93.0% (by HPLC)

Supplier: TCI America

B4598-500MG B4598-5G
TCB4598-500MGEA 154 USD
TCB4598-500MG TCB4598-5G
4,4'-Bis(dodecyloxy)-3-methylazobenzene ≥93.0% (by HPLC)
4,4'-Bis(dodecyloxy)-3-methylazobenzene
Formula: C₃₇H₆₀N₂O₂
CAS Number: 1440509-01-2

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 93.0 area%
Elemental analysis (Nitrogen) 4.50 to 5.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR