9,9-Dimethyl-2,7-bis[N-(m-tolyl)anilino]fluorene ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 9,9-Dimethyl-N,N'-diphenyl-N,N'-di(m-tolyl)fluorene-2,7-diamine, DMFL-TPD

D4920-1G D4920-200MG
TCD4920-1GEA 446 USD
TCD4920-1G TCD4920-200MG
9,9-Dimethyl-2,7-bis[N-(m-tolyl)anilino]fluorene ≥98.0% (by HPLC, total nitrogen)
9,9-Dimethyl-2,7-bis[N-(m-tolyl)anilino]fluorene
Formula: C₄₁H₃₆N₂
MDL Number: MFCD11112144
CAS Number: 143886-11-7

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR