3-(4-Chlorobutyl)indole-5-carbonitrile ≥97.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
3-(4-Chlorobutyl)-1H-indole-5-carbonitrile, 3-(4-Chlorobutyl)-5-cyanoindole
C3086-1G
C3086-5G
TCC3086-1GEA
57
USD
TCC3086-1G
TCC3086-5G
3-(4-Chlorobutyl)indole-5-carbonitrile ≥97.0% (by GC)
3-(4-Chlorobutyl)indole-5-carbonitrile
Formula:
C₁₃H₁₃ClN₂ MW: 232.71 g/mol |
MDL Number:
MFCD09833261 CAS Number: 143612-79-7 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 98.0 to 102.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...