n-Octylamine hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

O0484-1G O0484-5G
TCO0484-1GEA 48 USD
TCO0484-1G TCO0484-5G
n-Octylamine hydrochloride ≥98.0% (by titrimetric analysis)
n-Octylamine hydrochloride
Formula: C₈H₁₉N·HCl
Melting Pt: 198°C
MDL Number: MFCD00042019
CAS Number: 142-95-0
EINECS: 205-574-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR