6,6,7,7,8,8,8-Heptafluoro-5,5-bis(trifluoromethyl)-1,3-octadiene ≥98.0% (by GC)

Supplier: TCI America
Warning

H1515-25G H1515-5G
TCH1515-25GEA 479.1 USD
TCH1515-25G TCH1515-5G
6,6,7,7,8,8,8-Heptafluoro-5,5-bis(trifluoromethyl)-1,3-octadiene ≥98.0% (by GC)
6,6,7,7,8,8,8-Heptafluoro-5,5-bis(trifluoromethyl)-1,3-octadiene
Formula: C₁₀H₅F₁₃
MW: 372.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28014057
CAS Number: 1429188-77-1
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR