Dimethyl 2,2'-Bipyridine-6,6'-dicarboxylate ≥95.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 6,6'-Bis(methoxycarbonyl)-2,2'-bipyridine, 6,6'-Dimethoxycarbonyl-2,2'-bipyridine, 2,2'-Bipyridine-6,6'-dicarboxylic acid Dimethyl ester, Dimethyl [2,2'-bipyridine]-6,6'-dicarboxylate

D5025-1G D5025-200MG
TCD5025-1GEA 501 USD
TCD5025-1G TCD5025-200MG
Dimethyl 2,2'-Bipyridine-6,6'-dicarboxylate ≥95.0% (by GC)
Dimethyl 2,2'-Bipyridine-6,6'-dicarboxylate
Formula: C₁₄H₁₂N₂O₄
MW: 272.26 g/mol
MDL Number: MFCD11042478
CAS Number: 142593-07-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point gt. 192.0 and le 196.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR