3-Acetylpyridine N-Oxide ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
3-Acetylpyridine n-oxide
A2987-1G
A2987-5G
TCA2987-1GEA
80
USD
TCA2987-1G
TCA2987-5G
3-Acetylpyridine N-Oxide ≥98.0% (by GC)
3-Acetylpyridine N-Oxide
Formula:
C₇H₇NO₂ MW: 137.14 g/mol |
MDL Number:
MFCD00955798 CAS Number: 14188-94-4 |
Specification Test Results
Purity (GC) | min. 98.0 % |
Melting point | 144.0 to 148.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...