AquaMet ≥95.0% (by HPLC)
Supplier: TCI America
Synonyms:
[1,3-Bis(2,4,6-trimethylphenyl)-4-[(4-ethyl-4-methylpiperazinium-1-yl)methyl]-2-imidazolidinylidene]dichloro(2-isopropoxybenzylidene)ruthenium(II) chloride
A3009-100MG
A3009-500MG
76219-278EA
278
USD
76219-278
76219-280
AquaMet ≥95.0% (by HPLC)
AquaMet
Formula:
C₃₉H₅₅Cl₃N₄ORu MW: 803.32 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD28411647 CAS Number: 1414707-08-6 |
Specification Test Results
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Elemental analysis (Nitrogen) | 6.50 to 7.50 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...