9-Benzyl-2-bromo-9H-carbazole ≥98.0% (by GC, total nitrogen)

Supplier: TCI America

B5460-1G B5460-5G
TCB5460-1GEA 131 USD
TCB5460-1G TCB5460-5G
9-Benzyl-2-bromo-9H-carbazole ≥98.0% (by GC, total nitrogen)
9-Benzyl-2-bromo-9H-carbazole
Formula: C₁₉H₁₄BrN
MW: 336.23 g/mol
MDL Number: MFCD30469064
CAS Number: 1401863-51-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 133.0 to 137.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR