2,5-Bis[(trimethylsilyl)ethynyl]thieno[3,2-b]thiophene ≥98.0% (by GC)
Supplier: TCI America
B5196-1G
B5196-200MG
76219-518EA
596
USD
76219-518
76219-520
2,5-Bis[(trimethylsilyl)ethynyl]thieno[3,2-b]thiophene ≥98.0% (by GC)
2,5-Bis[(trimethylsilyl)ethynyl]thieno[3,2-b]thiophene
Formula:
C₁₆H₂₀S₂Si₂ MW: 332.63 g/mol |
CAS Number:
139896-65-4 |
Specification Test Results
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 211.0 to 215.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...