PDE5-IN-7 ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 5-(2-Ethoxyphenyl)-1-methyl-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-7(6H)-one, 5-(2-Ethoxyphenyl)-1-methyl-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]-7-pyrimidinone

E1134-25G E1134-5G
TCE1134-25GEA 234 USD
TCE1134-25G TCE1134-5G
PDE5-IN-7 ≥98.0% (by HPLC, titration analysis)
PDE5-IN-7
Formula: C₁₇H₂₀N₄O₂
MW: 312.37 g/mol
MDL Number: MFCD03093929
CAS Number: 139756-21-1

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 143.0 to 147.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR