1-Methyl-4-nitro-3-propylpyrazole-5-carboxamide ≥96.0% (by GC)

Supplier: TCI America
Warning

M2773-25G M2773-5G
76220-250EA 449 USD
76220-250 76220-252
1-Methyl-4-nitro-3-propylpyrazole-5-carboxamide ≥96.0% (by GC)
1-Methyl-4-nitro-3-propylpyrazole-5-carboxamide
Formula: C₈H₁₂N₄O₃
MDL Number: MFCD02253196
CAS Number: 139756-01-7
EINECS: 423-960-6

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 96.0 %
Melting point 137.0 to 141.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR