3,3'-Bi[1,4]benzoxazino[2,3,4-kl]phenoxazine

Supplier: TCI America

Synonyms: HN-D2

B4907-1G B4907-200MG
TCB4907-1GEA 883 USD
TCB4907-1G TCB4907-200MG
3,3'-Bi[1,4]benzoxazino[2,3,4-kl]phenoxazine
3,3'-Bi[1,4]benzoxazino[2,3,4-kl]phenoxazine
Formula: C₃₆H₂₀N₂O₄
MW: 544.57 g/mol
Melting Pt: 340°C
CAS Number: 1395881-58-9

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Elemental analysis (Nitrogen) 4.90 to 5.40 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR