Methyl 1-[(2'-Cyano-[1,1'-biphenyl]-4-yl)methyl]-2-ethoxy-1H-benzo[d]imidazole-7-carboxylate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

M3197-100G M3197-25G
TCM3197-100GEA 205 USD
TCM3197-100G TCM3197-25G
Methyl 1-[(2'-Cyano-[1,1'-biphenyl]-4-yl)methyl]-2-ethoxy-1H-benzo[d]imidazole-7-carboxylate ≥98.0% (by HPLC, total nitrogen)
Methyl 1-[(2'-Cyano-[1,1'-biphenyl]-4-yl)methyl]-2-ethoxy-1H-benzo[d]imidazole-7-carboxylate
Formula: C₂₅H₂₁N₃O₃
MDL Number: MFCD09031384
CAS Number: 139481-44-0

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 166.0 to 170.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR