5,5'-Diiodo-2,2'-dimethylbiphenyl ≥97.0% (by GC)
Supplier: TCI America
D5430-1G
D5430-5G
TCD5430-1GEA
174
USD
TCD5430-1G
TCD5430-5G
5,5'-Diiodo-2,2'-dimethylbiphenyl ≥97.0% (by GC)
5,5'-Diiodo-2,2'-dimethylbiphenyl
Formula:
C₁₄H₁₂I₂ |
CAS Number:
1392203-19-8 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 115.0 to 119.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...