1,5-Diethynyl-2,4-dimethylbenzene ≥98.0% (by GC)

Supplier: TCI America

D4878-1G D4878-5G
76219-732EA 175 USD
76219-732 76219-734
1,5-Diethynyl-2,4-dimethylbenzene ≥98.0% (by GC)
1,5-Diethynyl-2,4-dimethylbenzene
Formula: C₁₂H₁₀
CAS Number: 1379822-09-9

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Melting point 56.0 to 60.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR