Borane - Ammonia Complex ≥85.0% (V)

Supplier: TCI America

B5082-25G B5082-5G
TCB5082-25GEA 433.44 USD
TCB5082-25G TCB5082-5G
Borane - Ammonia Complex ≥85.0% (V)
Borane - Ammonia Complex
Formula: NH₃·BH₃
Storage Temperature: Refrigerator
MDL Number: MFCD00075639
CAS Number: 13774-81-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Gasburet method) min. 85.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR